Stampa…

O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-(6-cyano-2-methyl-7-oxo-4,8-dioxa-2,5-diazadec-5-en-3-ylidene)-N-methylmethanaminium tetrafluoroborate, TOTU

E0916-5G E0916-1G
TCIAE0916-5GEA 53 EUR
TCIAE0916-5G TCIAE0916-1G
O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate
Formula: C₁₀H₁₇BF₄N₄O₃
Peso molecolare: 328,07 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00192127
CAS Number: 136849-72-4
UN: 1759
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR