Stampa…

(-)-Epicatechin gallate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: (2R,3R)-2-(3,4-Dihydroxyphenyl)-5,7-dihydroxychroman-3-yl 3,4,5-trihydroxybenzoate, (-)-cis-3,3',4',5,7-Pentahydroxyflavane 3-Gallate

E0890-100MG E0890-20MG
TCIAE0890-100MGEA 350 EUR
TCIAE0890-100MG TCIAE0890-20MG
(-)-Epicatechin gallate ≥98.0% (by HPLC)
(-)-Epicatechin gallate
Formula: C₂₂H₁₈O₁₀
Peso molecolare: 442,38 g/mol
Punto di fusione: 255°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00075936
CAS Number: 1257-08-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -175.0 to -185.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR