Stampa…

Ethyl-1-methyl-3-piperidinecarboxylate ≥95.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Methylnipecotic Acid Ethyl Ester, Ethyl 1-Methylnipecotate, 1-Methyl-3-piperidinecarboxylic Acid Ethyl Ester, Ethyl N-methylpiperidine-3-carboxylate

E0863-25G E0863-5G
TCIAE0863-25GEA 113 EUR
TCIAE0863-25G TCIAE0863-5G
Ethyl-1-methyl-3-piperidinecarboxylate ≥95.0% (by GC, titration analysis)
Ethyl-1-methyl-3-piperidinecarboxylate
Formula: C₉H₁₇NO₂
Peso molecolare: 171,24 g/mol
Punto di ebollizione: 96°C
Flash Pt: 68°C
MDL Number: MFCD00006496
CAS Number: 5166-67-6
EINECS: 225-951-0

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR