Stampa…

Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: Ethyl 1-Benzyl-3-oxo-4-piperidinecarboxylate HCl, Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, Ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride

E0855-5G E0855-1G
TCIAE0855-5GEA 114 EUR
TCIAE0855-5G TCIAE0855-1G
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride ≥98.0% (by titrimetric analysis)
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride
Formula: C₁₅H₂₀ClNO₃
MDL Number: MFCD00012792
CAS Number: 52763-21-0
EINECS: 258-162-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR