N-Ethyl-o-crotonotoluidide ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(E)-N-Ethyl-N-(o-tolyl)but-2-enamide, N-Crotonyl-N-ethyl-o-toluidine, trans-N-Ethyl-N-(o-tolyl)-2-butenamide
E0814-25G
E0814-5G
TCIAE0814-25GEA
265
EUR
TCIAE0814-25G
TCIAE0814-5G
N-Ethyl-o-crotonotoluidide ≥95.0% (by GC)
N-Ethyl-o-crotonotoluidide
Formula:
C₁₃H₁₇NO Peso molecolare: 203,29 g/mol Punto di ebollizione: 155°C |
MDL Number:
MFCD00026989 CAS Number: 124236-29-9 EINECS: 207-596-3 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...