Stampa…

Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (2,6-Dichloro-5-fluoronicotinoyl)acetic Acid Ethyl Ester, Ethyl (2,6-Dichloro-5-fluoronicotinoyl)acetate, Ethyl 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionate, 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionic Acid Ethyl Ester

E0811-25G E0811-5G
TCIAE0811-25GEA 151 EUR
TCIAE0811-25G TCIAE0811-5G
Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate ≥98.0% (by titrimetric analysis)
Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate
Formula: C₁₀H₈Cl₂FNO₃
Peso molecolare: 280,08 g/mol
MDL Number: MFCD00799535
CAS Number: 96568-04-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 % (total of isomer)
Purity (Neutralization titration) min. 98.0 %
Melting point 66.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR