Stampa…

5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 5-Ethyl-3-hydroxy-4-methylfuran-2(5H)-one, Ethyl Fenugreek Lactone

E0800-25G E0800-5G
TCIAE0800-25GEA 209 EUR
TCIAE0800-25G TCIAE0800-5G
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone ≥97.0% (by GC, titration analysis)
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone
Formula: C₇H₁₀O₃
Peso molecolare: 142,15 g/mol
Punto di ebollizione: 94°C
MDL Number: MFCD00036673
CAS Number: 698-10-2
EINECS: 211-811-6

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Freezing point 25.0 to 29.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR