Stampa…

1-Ethyl-3-methylimidazolium nitrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

E0775-5G E0775-25G
TCIAE0775-5GEA 62 EUR
TCIAE0775-5G TCIAE0775-25G
1-Ethyl-3-methylimidazolium nitrate ≥98.0% (by HPLC, titration analysis)
1-Ethyl-3-methylimidazolium nitrate
Formula: C₆H₁₁N₃O₃
Peso molecolare: 173,17 g/mol
Punto di fusione: 41°C
MDL Number: MFCD00210007
CAS Number: 143314-14-1
UN: 1479
ADR: 5.1,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR