Stampa…

Enoxacin sesquihydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 1-Ethyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid sesquihydrate, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic Acid

E0762-25G E0762-5G
TCIAE0762-25GEA 212 EUR
TCIAE0762-25G TCIAE0762-5G
Enoxacin sesquihydrate ≥98.0% (by HPLC, titration analysis)
Enoxacin sesquihydrate
Formula: C₃₀H₄₀F₂N₈O₉
Peso molecolare: 694,68 g/mol
MDL Number: MFCD01747755
CAS Number: 84294-96-2
Merck Index: 14,03587

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Melting point 225.0 to 229.0 deg-C
Drying loss 7.0 to 9.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR