Ethyl-4-hydroxy-3-methoxycinnamate ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
4-Hydroxy-3-methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-hydroxy-3-methoxyphenyl)acrylate, Ethyl Ferulate, Ferulic Acid Ethyl Ester
Formula:
C₁₂H₁₄O₄ Peso molecolare: 222.24 g/mol Punto di ebollizione: 185°C |
MDL Number:
MFCD00009190 CAS Number: 4046-02-0 EINECS: 223-745-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 56.0 to 61.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...