Stampa…

Ethyl-4-chloro-1-piperidinecarboxylate ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 4-Chloro-1-(ethoxycarbonyl)piperidine, 1-Carbethoxy-4-chloropiperidine, 4-Chloro-1-piperidinecarboxylic Acid Ethyl Ester, Ethyl 4-chloropiperidine-1-carboxylate

E0732-25G E0732-5G
TCIAE0732-25GEA 103 EUR
TCIAE0732-25G TCIAE0732-5G
Ethyl-4-chloro-1-piperidinecarboxylate ≥98.0% (by GC)
Ethyl-4-chloro-1-piperidinecarboxylate
Formula: C₈H₁₄ClNO₂
Flash Pt: 108°C
MDL Number: MFCD09038524
CAS Number: 152820-13-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR