Stampa…

Ethyl-5-chloro-1H-indole-2-carboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 5-chloro-1H-indole-2-carboxylate, Ethyl 5-chloro-2-indolecarboxylate

E0696-25G E0696-5G
TCIAE0696-25GEA 201 EUR
TCIAE0696-25G TCIAE0696-5G
Ethyl-5-chloro-1H-indole-2-carboxylate ≥98.0% (by GC)
Ethyl-5-chloro-1H-indole-2-carboxylate
Formula: C₁₁H₁₀ClNO₂
Peso molecolare: 223,66 g/mol
MDL Number: MFCD00005610
CAS Number: 4792-67-0
EINECS: 225-345-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 168.0 to 172.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR