Ethyl stilbene-4-carboxylate ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
Stilbene-4-carboxylic Acid Ethyl Ester, 4-(Ethoxycarbonyl)stilbene
Formula:
C₁₇H₁₆O₂ |
MDL Number:
MFCD06204340 CAS Number: 109463-48-1 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...