Stampa…

Ethyl-4-cyanocinnamate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Cyanocinnamic Acid Ethyl Ester

E0677-1G E0677-5G
TCIAE0677-1GEA 60 EUR
TCIAE0677-1G TCIAE0677-5G
Ethyl-4-cyanocinnamate ≥98.0% (by GC)
Ethyl-4-cyanocinnamate
Formula: C₁₂H₁₁NO₂
Peso molecolare: 201,23 g/mol
CAS Number: 62174-99-6
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR