Etoposide ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(5R,5aR,8aR,9S)-9-(((4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-methylhexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one, 4'-Demethylepipodophyllotoxin 9-(4,6-O-Ethylidene-beta-D-glucopyranoside)
Formula:
C₂₉H₃₂O₁₃ Peso molecolare: 588,56 g/mol Punto di fusione: 251°C |
MDL Number:
MFCD00869325 CAS Number: 33419-42-0 EINECS: 251-509-1 Merck Index: 14,03886 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Water | max. 7.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...