Stampa…
3,3'-(Ethylenedithio)dipropionitrile ≥97.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 1,2-Bis(2-cyanoethylthio)ethane, 4,7-Dithiadecanedinitrile
E0673-1G E0673-5G
TCIAE0673-1GEA 68 EUR
TCIAE0673-1G TCIAE0673-5G
3,3'-(Ethylenedithio)dipropionitrile ≥97.0% (by GC)
3,3'-(Ethylenedithio)dipropionitrile
Formula: C₈H₁₂N₂S₂
Peso molecolare: 200,32 g/mol
CAS Number: 86180-54-3
Visualizza i codici
Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 91.0 to 95.0 deg-C