Ethyl-2-fluoropropionate ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Fluoropropionic Acid Ethyl Ester, Ethyl 2-fluoropropionate
E0663-25G
E0663-5G
TCIAE0663-25GEA
66
EUR
TCIAE0663-25G
TCIAE0663-5G
Ethyl-2-fluoropropionate ≥97.0% (by GC)
Ethyl-2-fluoropropionate
Formula:
C₅H₉FO₂ Peso molecolare: 120,12 g/mol Punto di ebollizione: 121°C |
MDL Number:
MFCD01320801 CAS Number: 349-43-9 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...