Stampa…

Ethyl-4-(Chloromethyl)benzoate ≥98.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: 4-(Chloromethyl)benzoic Acid Ethyl Ester, Ethyl 4-(chloromethyl)benzoate

E0662-25G E0662-5G
TCIAE0662-25GEA 136 EUR
TCIAE0662-25G TCIAE0662-5G
Ethyl-4-(Chloromethyl)benzoate ≥98.0% (by GC)
Ethyl-4-(Chloromethyl)benzoate
Formula: C₁₀H₁₁ClO₂
Peso molecolare: 198.65 g/mol
Punto di ebollizione: 280°C
MDL Number: MFCD04337336
CAS Number: 1201-90-7
UN: 1760
ADR: 8 III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR