Ethyl-3-(chloromethyl)benzoate ≥96.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Ethyl 3-(chloromethyl)benzoate, 3-(Chloromethyl)benzoic Acid Ethyl Ester
Formula:
C₁₀H₁₁ClO₂ Peso molecolare: 198.65 g/mol Punto di ebollizione: 169°C |
MDL Number:
MFCD06200832 CAS Number: 54589-54-7 UN: 3265 ADR: 8 III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...