Stampa…

Ethyl-4-acetamidophenylacetate ≥98.0% (by GC)

Fornitore: TCI

E0585-25G E0585-5G
TCIAE0585-25GEA 116 EUR
TCIAE0585-25G TCIAE0585-5G
Ethyl-4-acetamidophenylacetate ≥98.0% (by GC)
Ethyl-4-acetamidophenylacetate
Formula: C₁₂H₁₅NO₃
MDL Number: MFCD06797104
CAS Number: 13475-17-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR