Stampa…

2-(5-Ethylpyridin-2-yl)ethanol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

E0562-25G E0562-5G
TCIAE0562-25GEA 86 EUR
TCIAE0562-25G TCIAE0562-5G
2-(5-Ethylpyridin-2-yl)ethanol ≥98.0% (by GC)
2-(5-Ethylpyridin-2-yl)ethanol
Formula: C₉H₁₃NO
Peso molecolare: 151,21 g/mol
MDL Number: MFCD00129040
CAS Number: 5223-06-3
EINECS: 226-024-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR