N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, NEPA, N-[(S)-1-Carboxyethyl]-L-homophenylalanine Ethyl Ester, (S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanoic acid, N-(1S)-Ethoxycarbonyl-3-phenylpropyl-(S)-alanine
E0530-25G
E0530-5G
TCIAE0530-25GEA
79
EUR
TCIAE0530-25G
TCIAE0530-5G
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine ≥98.0% (by HPLC, titration analysis)
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
Formula:
C₁₅H₂₁NO₄ Peso molecolare: 279,34 g/mol |
MDL Number:
MFCD00209976 CAS Number: 82717-96-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 148.0 to 154.0 deg-C |
Specific rotation [a]20/D | +28.0 to +31.0 deg (C=1, Methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...