Ethyl nonafluorobutyl ether (mixture of isomers) ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Perfluorobutyl Ethyl Ether, Nonafluorobutyl Ethyl Ether, Ethyl Perfluorobutyl Ether
E0528-25G
E0528-500G
TCIAE0528-25GEA
26
EUR
TCIAE0528-25G
TCIAE0528-500G
Ethyl nonafluorobutyl ether (mixture of isomers) ≥98.0% (by GC)
Ethyl nonafluorobutyl ether (mixture of isomers)
Formula:
C₆H₅F₉O Peso molecolare: 264,09 g/mol |
CAS Number:
813458-04-7 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Specific gravity (20/20) | 1.4350 to 1.4450 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...