Ethylene glycol monovinyl ether ≥98.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Vinyl Cellosolve, Ethylene glycol vinyl ether, 2-Vinyloxyethanol, Vinyl Glycol, 2-(Vinyloxy)ethan-1-ol
Stabilization:
Stabilised with potassium hydroxide
E0518-25ML
E0518-500ML
TCIAE0518-25MLEA
25
EUR
TCIAE0518-25ML
TCIAE0518-500ML
Ethylene glycol monovinyl ether ≥98.0% (by GC) stabilizzato
Ethylene glycol monovinyl ether
Formula:
C₄H₈O₂ Peso molecolare: 88,11 g/mol Punto di ebollizione: 143°C Flash Pt: 49°C |
MDL Number:
MFCD00192151 CAS Number: 764-48-7 EINECS: 212-124-4 UN: 3271 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...