Stampa…

Emodin ≥96.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,3,8-Trihydroxy-6-methylanthracene-9,10-dione, 1,3,8-Trihydroxy-6-methylanthraquinone

E0500-1G E0500-100MG
TCIAE0500-1GEA 380 EUR
TCIAE0500-1G TCIAE0500-100MG
Emodin ≥96.0% (by HPLC)
Emodin
Formula: C₁₅H₁₀O₅
Peso molecolare: 270,24 g/mol
MDL Number: MFCD00001207
CAS Number: 518-82-1
EINECS: 208-258-8
Merck Index: 14,03561

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 96.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR