Stampa…

Ethyl-N-phenylformimidate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: N-Phenylformimidic Acid Ethyl Ester, Ethyl N-phenylformimidate, N-Ethoxymethyleneaniline

E0495-25G E0495-5G
TCIAE0495-25GEA 66 EUR
TCIAE0495-25G TCIAE0495-5G
Ethyl-N-phenylformimidate ≥98.0% (by GC)
Ethyl-N-phenylformimidate
Formula: C₉H₁₁NO
Peso molecolare: 149,19 g/mol
Punto di ebollizione: 215°C
MDL Number: MFCD00043655
CAS Number: 6780-49-0
EINECS: 229-841-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR