Stampa…

Ethylenediaminetetraacetic dianhydride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: EDTA Dianhydride, 4,4'-Ethylenebis(2,6-morpholinedione), 4,4-(Ethane-1,2-diyl)bis(morpholine-2,6-dione)

E0480-25G E0480-100G
TCIAE0480-25GEA 108 EUR
TCIAE0480-25G TCIAE0480-100G
Ethylenediaminetetraacetic dianhydride ≥98.0% (by titrimetric analysis)
Ethylenediaminetetraacetic dianhydride
Formula: C₁₀H₁₂N₂O₆
Peso molecolare: 256,21 g/mol
MDL Number: MFCD00074963
CAS Number: 23911-25-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR