Ethylene di(thiotosylate) ≥98.0% (by HPLC) protecting reagent for active methylene
Fornitore: TCI
Formula:
C₁₆H₁₈O₄S₄ |
MDL Number:
MFCD00008550 CAS Number: 2225-23-2 EINECS: 218-752-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 72.0 to 75.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...