Ethynyl-p-tolyl sulphone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Tosylacetylene, Ethynyl p-Tolyl sulphone, p-Toluenesulphonylacetylene
E0466-1G
E0466-5G
TCIAE0466-1GEA
156
EUR
TCIAE0466-1G
TCIAE0466-5G
Ethynyl-p-tolyl sulphone ≥98.0% (by GC)
Ethynyl-p-tolyl sulphone
Formula:
C₉H₈O₂S Peso molecolare: 180,22 g/mol |
MDL Number:
MFCD00191647 CAS Number: 13894-21-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 73.0 to 77.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...