Stampa…

D-Erythronolactone ≥98.0% (by GC)

Fornitore: TCI

E0455-1G E0455-25G E0455-5G
TCIAE0455-1GEA 39 EUR
TCIAE0455-1G TCIAE0455-25G TCIAE0455-5G
D-Erythronolactone ≥98.0% (by GC)
D-Erythronolactone
Formula: C₄H₆O₄
MDL Number: MFCD00077763
CAS Number: 15667-21-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C
Specific rotation [a]20/D -69.0 to -75.0 deg (C=1, H2O)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR