Stampa…

Ethyl (S)-(-)-1-ethyl-2-pyrrolidinecarboxylate ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

E0450-1G E0450-5G
TCIAE0450-1GEA 39 EUR
TCIAE0450-1G TCIAE0450-5G
Ethyl (S)-(-)-1-ethyl-2-pyrrolidinecarboxylate ≥98.0% (by GC, titration analysis)
Ethyl (S)-(-)-1-ethyl-2-pyrrolidinecarboxylate
Formula: C₉H₁₇NO₂
MDL Number: MFCD00191434
CAS Number: 938-54-5

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -90.0 to -98.0 deg (C=0.6, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR