Ethyl Methyl Ketone 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Methyl Ethyl Ketone 2,4-Dinitrophenylhydrazone, 2-Butanone 2,4-Dinitrophenylhydrazone
Formula:
C₁₀H₁₂N₄O₄ Peso molecolare: 252.23 g/mol |
MDL Number:
MFCD00191644 CAS Number: 958-60-1 UN: 1325 ADR: 4.1 III |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...