N-Carbethoxy-4-piperidone ≥97.0% (by GC)
Fornitore: TCI
Synonyms:
4-Piperidone-N-carboxylic acid ethyl ester, N-Ethoxycarbonyl-4-piperidone, Ethyl4-oxo-1-piperidinecarboxylate
Formula:
C₈H₁₃NO₃ Punto di ebollizione: 151°C Flash Pt: 87°C |
MDL Number:
MFCD00006188 CAS Number: 29976-53-2 EINECS: 249-984-5 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...