Stampa…

exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Anhydride, exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic Anhydride

E0392-25G E0392-100G
TCIAE0392-25GEA 40 EUR
TCIAE0392-25G TCIAE0392-100G
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride ≥98.0% (by titrimetric analysis)
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride
Formula: C₈H₆O₄
Peso molecolare: 166,13 g/mol
MDL Number: MFCD00151506
CAS Number: 6118-51-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Methanolysis Method) min. 98.0 %
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR