1-Ethyl-4-(methoxycarbonyl)pyridinium iodide ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Carbomethoxy-1-ethylpyridinium Iodide
E0362-1G
E0362-5G
TCIAE0362-1GEA
25
EUR
TCIAE0362-1G
TCIAE0362-5G
1-Ethyl-4-(methoxycarbonyl)pyridinium iodide ≥97.0% (by titrimetric analysis)
1-Ethyl-4-(methoxycarbonyl)pyridinium iodide
Formula:
C₉H₁₂INO₂ Peso molecolare: 293,10 g/mol |
MDL Number:
MFCD00011996 CAS Number: 1199-65-1 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (Potassium iodate Method) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 109.0 to 113.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...