Ethylene trithiocarbonate ≥96.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
1,3-Dithiolane-2-thione
E0323-25G
E0323-100G
TCIAE0323-25GEA
137
EUR
TCIAE0323-25G
TCIAE0323-100G
Ethylene trithiocarbonate ≥96.0% (by GC)
Ethylene trithiocarbonate
Formula:
C₃H₄S₃ Peso molecolare: 136,25 g/mol |
MDL Number:
MFCD00005493 CAS Number: 822-38-8 EINECS: 212-498-9 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...