Stampa…

Ethylenediamine-N,N'-diacetic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: Ethylene-N,N'-diglycine, EDDA

E0275-25G E0275-5G
TCIAE0275-25GEA 213 EUR
TCIAE0275-25G TCIAE0275-5G
Ethylenediamine-N,N'-diacetic acid ≥98.0% (by titrimetric analysis)
Ethylenediamine-N,N'-diacetic acid
Formula: C₆H₁₂N₂O₄
Peso molecolare: 176.17 g/mol
MDL Number: MFCD00004281
CAS Number: 5657-17-0
EINECS: 227-105-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR