5-Ethylidene-2-norbornene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
5-Ethylidenebicyclo[2.2.1]hept-2-ene
E0244-25ML
E0244-500ML
TCIAE0244-25MLEA
17
EUR
TCIAE0244-25ML
TCIAE0244-500ML
5-Ethylidene-2-norbornene ≥98.0% (by GC)
5-Ethylidene-2-norbornene
Formula:
C₉H₁₂ Peso molecolare: 120,20 g/mol Punto di ebollizione: 147°C Flash Pt: 38°C |
MDL Number:
MFCD00167576 CAS Number: 16219-75-3 EINECS: 240-347-7 UN: 3295 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...