Stampa…

Ethyl Isopropyl sulphide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

E0225-10G E0225-5G E0225-25G
TCIAE0225-10GEA 79 EUR
TCIAE0225-10G TCIAE0225-5G TCIAE0225-25G
Ethyl Isopropyl sulphide ≥98.0% (by GC)
Ethyl Isopropyl sulphide
Formula: C₅H₁₂S
Peso molecolare: 104,21 g/mol
Punto di ebollizione: 103°C
MDL Number: MFCD00039908
CAS Number: 5145-99-3
EINECS: 225-917-5
UN: 1993
ADR: 3,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR