Ethylene glycol dibutyl ether ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Ethyleneglycoldi-n-butylether, Dibutyl Glycol, Dibutyl Cellosolve, 1,2-Dibutoxyethane
E0217-25ML
E0217-500ML
TCIAE0217-25MLEA
25
EUR
TCIAE0217-25ML
TCIAE0217-500ML
Ethylene glycol dibutyl ether ≥98.0% (by GC)
Ethylene glycol dibutyl ether
Formula:
C₁₀H₂₂O₂ Peso molecolare: 174,28 g/mol Punto di ebollizione: 202°C Punto di fusione: -69°C Flash Pt: 85°C |
MDL Number:
MFCD00048785 CAS Number: 112-48-1 EINECS: 203-976-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...