Stampa…

N-Ethyl-p-toluidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-(Ethylamino)toluene, N-Ethyl-4-methylaniline

E0179-25ML E0179-100ML
TCIAE0179-25MLEA 68 EUR
TCIAE0179-25ML TCIAE0179-100ML
N-Ethyl-p-toluidine ≥98.0% (by GC, titration analysis)
N-Ethyl-p-toluidine
Formula: C₉H₁₃N
Peso molecolare: 135,21 g/mol
Punto di ebollizione: 222°C
Flash Pt: 89°C
MDL Number: MFCD00035793
CAS Number: 622-57-1
EINECS: 210-742-9
UN: 2754
ADR: 6.1,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR