Stampa…

Ethylenediaminetetraacetic acid disodium zinc salt ≥98.0% (by titrimetric analysis)

Fornitore: TCI

E0098-25G E0098-500G
TCIAE0098-25GEA 22 EUR
TCIAE0098-25G TCIAE0098-500G
Ethylenediaminetetraacetic acid disodium zinc salt ≥98.0% (by titrimetric analysis)
Ethylenediaminetetraacetic acid disodium zinc salt
Formula: C₁₀H₁₂N₂Na₂O₈Zn
MDL Number: MFCD00135187
CAS Number: 14025-21-9
EINECS: 237-865-0

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR