Stampa…

Ethylenediaminetetraacetic acid lead(II) disodium salt hydrate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Lead(II) Disodium Ethylenediaminetetraacetate

E0093-25G E0093-500G
TCIAE0093-25GEA 38 EUR
TCIAE0093-25G TCIAE0093-500G
Ethylenediaminetetraacetic acid lead(II) disodium salt hydrate ≥98.0% (by titrimetric analysis)
Ethylenediaminetetraacetic acid lead(II) disodium salt hydrate
Formula: C₁₀H₁₆N₂O₈
Peso molecolare: 541,39 g/mol
MDL Number: MFCD00148921
CAS Number: 22904-40-1
EINECS: 245-314-0
UN: 2291
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 6.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR