1,3-Diidrossiacetone ≥97.0% (by GC)
Fornitore: TCI
D5818-25G
D5818-5G
TCIAD5818-25GEA
43
EUR
TCIAD5818-25G
TCIAD5818-5G
1,3-Diidrossiacetone ≥97.0% (by GC)
1,3-Diidrossiacetone
Formula:
C₃H₆O₃ |
MDL Number:
MFCD00004670 CAS Number: 96-26-4 EINECS: 202-494-5 Merck Index: 14,03182 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...