2,4-Dimethoxy-3-methylbenzaldehyde ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₂O₃ Punto di ebollizione: 109°C |
MDL Number:
MFCD00011489 CAS Number: 7149-92-0 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...