Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III) ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
[(ppy)2IrCl]2, Dichlorotetrakis[2-(2-pyridinyl)phenyl]diiridium(III), Dichlorotetrakis(2-(2-pyridinyl)phenyl)diiridium(III)
D5744-1G
D5744-200MG
TCIAD5744-1GEA
363
EUR
TCIAD5744-1G
TCIAD5744-200MG
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III) ≥98.0% (by HPLC)
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)
Formula:
C₄₄H₃₂Cl₂Ir₂N₄ Peso molecolare: 1072,09 g/mol |
MDL Number:
MFCD07784515 CAS Number: 92220-65-0 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...