2,4-Dimethyl-3-cyclohexene-1-carboxaldehyde (mixture of isomers) ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₉H₁₄O Punto di ebollizione: 189°C |
MDL Number:
MFCD00221550 CAS Number: 68039-49-6 EINECS: 268-264-1 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...