Stampa…

2,6-Dichloroquinoxaline ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D5697-5G D5697-25G
TCIAD5697-5GEA 54 EUR
TCIAD5697-5G TCIAD5697-25G
2,6-Dichloroquinoxaline ≥98.0% (by GC)
2,6-Dichloroquinoxaline
Formula: C₈H₄Cl₂N₂
Peso molecolare: 199,0368 g/mol
MDL Number: MFCD00839695
CAS Number: 18671-97-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR