Ditrimethylolpropane tetraacrylate ≥75.0% (by GC) stabilizzato
Fornitore: TCI
|
Attenzione
|
Synonyms:
[Oxybis(methylene)]bis(2-ethylpropane-3,2,1-triyl) tetraacrylate, Di(Trimethylolpropane)tetraacrylate, Acrylic acid [Oxybis(methylene)]bis(2-ethylpropane-3,2,1-triyl) tetraester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
Formula:
C₂₄H₃₄O₉ |
MDL Number:
MFCD00192118 CAS Number: 94108-97-1 EINECS: 302-434-9 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 75.0 % |
Specific gravity (20/20) | 1.1050 to 1.1150 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...