1-(2,6-Diethylphenyl)-2,2,4-trimethyl-4-phenyl-3,4-dihydro-2H-pyrrol-1-ium tetrafluoroborate ≥98.0% (by HPLC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₂₃H₃₀BF₄N |
CAS Number:
1671098-46-6 UN: 1759 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...